missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ GM-CSF Antibody (namilumab), Novus Biologicals™

Human Monoclonal Antibody

Marque:  Novus Biologicals™ NBP3-28668-50ug

Code nomenclature Nacres: NA.46

 Afficher plus de versions de ce produit

Code produit. 30047481

  • 213.00€ / 50µg

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

GM-CSF Monoclonal antibody specifically detects GM-CSF in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Spécification

Spécification

GM-CSF
Monoclonal
Unconjugated
P04141
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
namilumab
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
CSF2/ GM-CSF
50 μg
Biologically Active Proteins, Cytokine Research, Immunology
1437
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.