Learn More
GNL3 Mouse anti-Human, Unconjugated, Clone: 3B8F10, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67169-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The GNL3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GNL3. This antibody recognizes human, mouse, rat antigen. The GNL3 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Spécification
| GNL3 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunofluorescence, ELISA | |
| 3B8F10 | |
| Unconjugated | |
| 20 μL | |
| 26354 | |
| IgG2a |