missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GORASP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10598-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
GRASP55 is a PDZ containing protein involved in the stacking of golgi cisternea in a cell-free system.Spécification
GORASP2 | |
Polyclonal | |
Unconjugated | |
GORASP2 | |
Golgi phosphoprotein 6, GOLPH6, GORASP2, GRASP55, GRS2, p59 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
113961, 26003, 70231 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q99JX3, Q9H8Y8, Q9R064 | |
Gorasp2 | |
GORASP2 Fusion Protein Ag0950 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |