Learn More
GPD1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 27943-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16835915
  • 399.00€ / 150µL
Expédition estimée: 07-06-2024
pour voir le stock disponible 
Description
Description
The GPD1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GPD1. This antibody recognizes human, mouse, rat antigen. The GPD1 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
GPD1
Polyclonal
Unconjugated
150 μL
2819
IgG
Western Blot, ELISA
333 μg/mL
Rabbit
Primary
Human, Rat, Mouse