missing translation for 'onlineSavingsMsg'
Learn More

GPR21 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 17973-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16827994

  • 325.38€ / 150µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Human follicle stimulating hormone is a glycoprotein produced from urine of post-menopausal women and having a total molecular mass of 35,500 Dalton. HuFSH is a heterodimeric hormone consisting of 165 amino acids a chain (non-active) and 115 amino acids b chain (active). HuFSH induces maturation of Graafian follicles of the ovary, promotes development of germinal cells in males and activates cytosolic tyrosine kinase. The HuFSH is purified by proprietary chromatographic techniques.
Spécification

Spécification

GPR21
Polyclonal
Unconjugated
GPR21
G protein coupled receptor 21, GPR21
Rabbit
Antigen Affinity Chromatography
RUO
2844, 338346, 689917
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BX79, Q99679
GPR21
GPR21 Fusion Protein Ag12064
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions