Learn More
GPR4 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28232-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The GPR4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GPR4. This antibody recognizes human, mouse antigen. The GPR4 antibody has been validated for the following applications: IHC, ELISA analysis.
Spécification
| GPR4 | |
| 550 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Immunohistochemistry, ELISA | |
| G protein coupled receptor 19, G protein coupled receptor 4, GPR19, GPR4 | |
| 20 μL | |
| 2828 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |