missing translation for 'onlineSavingsMsg'
Learn More

GPR81 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20146-1-AP-150UL

Nacres classification code: NA.46

 View more versions of this product

Product Code. 16829654

  • 410.00€ / 150µL

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Description

Description

The G protein-coupled receptor GPCR GPR81 has been reported primarily in adipose and pituitary. ESTs have been isolated from human bladder cancer and colon cancer libraries. It has not been detected in frontal, temporal and occipital lobes of the cortex, basal forebrain, caudate nucleus, nucleus accumbens, and hippocampus.
Specifications

Specifications

GPR81
Polyclonal
Unconjugated
HCAR1
FKSG80, G protein coupled receptor 104, G protein coupled receptor 81, GPR104, GPR81, LACR1, TA GPCR
Rabbit
Antigen Affinity Chromatography
RUO
27198
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BXC0
HCAR1
GPR81 Fusion Protein Ag13682
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.