missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16757-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
16757-1-AP antibody recognizes two different isoforms of GPT2 protein.
GPT and GPT2 (EC 2.6.1.2), also known as alanine transaminases, are pyridoxal enzymes that catalyze the reversible transamination between alanine and 2-oxoglutarate to form pyruvate and glutamate. By mediating the conversion of these 4 major intermediate metabolites, these transaminases have roles in gluconeogenesis and in amino acid metabolism.Spécification
GPT2 | |
Polyclonal | |
Unconjugated | |
GPT2 | |
AAT2, Alanine aminotransferase 2, ALT2, GPT 2, GPT2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
108682, 307759, 84706 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BGT5, Q8TD30 | |
GPT2 | |
GPT2 Fusion Protein Ag10273 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |