missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPX8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16846-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
GPX8 (Probable glutathione peroxidase 8) is also named as GSHPx-8 and belongs to the glutathione peroxidase family. Glutathione peroxidase (GPx) reduces hydroperoxides, including hydrogen peroxides, in the presence of reduced glutathione as a means of protecting organisms from oxidative damage. Several GPx isozymes have been identified in animal cells and these have been classified into different groups according to their cellular location and substrate specificity. In humans, eight types of GPx have been identified from GPx1 to GPx8. The full length GPX8 has 209 amino acids and the molecular weight is 24 kDa.Spécification
GPX8 | |
Polyclonal | |
Unconjugated | |
GPX8 | |
GPx 8, GPX8, GSHPx 8, UNQ847 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
294744, 493869, 69590 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TED1, Q9D7B7 | |
GPX8 | |
GPX8 Fusion Protein Ag9930 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |