missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK-3 beta Antibody (3D10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Marque: Novus Biologicals NBP1-47470
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
GSK-3 beta Monoclonal specifically detects GSK-3 beta in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spécification
| GSK-3 beta | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:2000, Simple Western 1:50, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK-3 beta, GSK3beta isoform | |
| Mouse | |
| 46 kDa | |
| 0.1 mL | |
| Cancer, Hypoxia, Lipid and Metabolism, mTOR Pathway, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| 2932 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 3D10 | |
| Unconjugated | |
| P49841 | |
| GSK3B | |
| Purified recombinant fragment of human GSK3 beta expressed in E. coli. [UniProt# P49841] | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Primate | |
| Precipitation |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu