missing translation for 'onlineSavingsMsg'
Learn More

GSNOR/ADH5 Mouse anti-Human, Clone: 5E6E2, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 66193-1-IG-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837845

  • 325.38€ / 150µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. The encoded protein forms a homodimer. It has virtually no activity for ethanol oxidation, but exhibits high activity for oxidation of long-chain primary alcohols and for oxidation of S-hydroxymethyl-glutathione, a spontaneous adduct between formaldehyde and glutathione. This enzyme is an important component of cellular metabolism for the elimination of formaldehyde, a potent irritant and sensitizing agent that causes lacrymation, rhinitis, pharyngitis, and contact dermatitis. The human genome contains several non-transcribed pseudogenes related to this gene.
Spécification

Spécification

GSNOR/ADH5
Monoclonal
0.76 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P11766
ADH5
GSNOR,ADH5 Fusion Protein Ag9511
150 μL
Primary
Human
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
5E6E2
Unconjugated
ADH5
ADH 3, ADH5, ADHX, Alcohol dehydrogenase 5, Alcohol dehydrogenase class 3, FALDH, FDH, GSH FDH, GSNOR, GSNOR,ADH5
Mouse
Protein G
RUO
128
-20°C
Liquid
Vidéos
FDS
Documents
Certificats
Promotions

Promotions