missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GYG2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13150-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
GYG2 belongs to the glycogenin family. Glycogenin is a self-glycosylating protein involved in the initiation phase of glycogen biosynthesis. Multiple alternatively spliced transcript variants encoding different isoforms have been identified, including the predomit band around 66kDa and a light band around 45kDa.Spécification
GYG2 | |
Polyclonal | |
Unconjugated | |
GYG2 | |
glycogenin 2, GN 2, GN2, GYG2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8908 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15488 | |
GYG2 | |
GYG2 Fusion Protein Ag3765 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |