Learn More
H-2Kb Monoclonal Antibody (AF6-88.5), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA516562
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Mouse MHC Class I H-2Kb antibody, clone AF6-88.5 recognizes the H-2Kb determit present on all nucleated cells of mice expressing this MHC class I haplotype.
H-2KbDb monoclonal antibody is specific for cells expressing the H-2K antigen coded for by the b haplotype, and for cells expressing the H-2D antigen coded for by the b haplotype.Spécification
H-2Kb | |
Monoclonal | |
0.5 mg/mL | |
H2-K1 | |
H-2K, H-2K(d), H2-K, K-f,H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H-2K(B); H-2K(K); H-2K(Q); MHC H2-K transplantation antigen; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I antigen; MHC class I heavy chain H2-K; class I major histocom | |
Mouse | |
Affinity chromatography | |
RUO | |
14972 | |
4°C or -20°C if preferred | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
AF6-88.5 | |
Unconjugated | |
P01902 | |
H2-K1 | |
C57BL mouse splenocytes | |
250 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |