Learn More
HAI-2/SPINT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Marque: Novus Biologicals NBP1-83299
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
HAI-2/SPINT2 Polyclonal specifically detects HAI-2/SPINT2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Spécification
| Immunohistochemistry (Paraffin) | |
| Rabbit | |
| RUO |
| Polyclonal | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:CLVSKVVGRCRASMPRWWYNVTDGSCQLFVYGGCDGNSNNYLTKEECLKKCATVTENATGDLATSRNAADSSVPSAPRRQDSEDHSSDMFNYEEYCTANAVTGPCRASFPRWYFDVERNSCNNFIYGGCRGNKNSYRSE | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |