Learn More
HBcAg Antibody (H50) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marque:  Novus Biologicals NB100-64452-0.025MG
Code nomenclature Nacres: NA.46
Code produit. 18093616
  • 204.00€ / 25µg
Description
HBcAg Monoclonal specifically detects HBcAg in Virus samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
HBcAg
Monoclonal
0.5 mg/mL
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000
c, Core antigen, HBc, HBcAg, Hepatitis B Virus core antigen
Mouse
Protein A or G purified
RUO
Primary
Recognises the core antigen.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Immunofluorescence, Immunohistochemistry, Immunocytochemistry
H50
Unconjugated
Phosphate buffered saline with 0.05% Sodium Azide
C
Native
0.025 mg
Virology Bacteria and Parasites
944568
Virus
Purified
For Research Use Only