Learn More
HDAC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16160-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody is a rabbit polyclonal antibody raised against a peptide mapping near the N-terminus of human HDAC1 and is specific to HDAC1. It does not react with other HDACs.
Transcriptional coactivator for CREB1 which activates transcription through both consensus and variant cAMP response element (CRE) sites.Acts as a coactivator, in the SIK/TORC signaling pathway,being active when dephosphorylated and acts independently of CREB1 ′Ser-133′ phosphorylation.Enhances the interaction of CREB1 with TAF4.Regulates the expression of specific CREB-activated genes such as the steroidogenic gene,StAR.Potent coactivator of PGC1alpha and inducer of mitochondrial biogenesis in muscle cells.Also coactivator for TAX activation of the human T-cell leukemia virus type 1 (HTLV-1) long terminal repeats (LTR).In the hippocampus, involved in late-phase long-term potentiation (L-LTP) maintece at the Schaffer collateral-CA1 synapses.May be required for dendritic growth of developing cortical neurons.Spécification
HDAC1 | |
Polyclonal | |
Unconjugated | |
HDAC1 | |
GON 10, HD1, HDAC1, histone deacetylase 1, RPD3, RPD3L1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
297893, 3065, 433759 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O09106, Q13547, Q4QQW4 | |
Hdac1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |