Learn More
HEXA Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 29259-1-AP-150UL
Informations supplémentaires : Poids : 0.00020kg
Code produit. 17261436
  • 399.00€ / 150µL
Expédition estimée: 24-05-2024
Description
Beta-hexosaminidase subunit alpha is involved in catalysis of degradation of the ganglioside GM2. Disfunction of beta-hexosaminidase leads to an accumulation of GM2 ganglioside in neurons and neurodegenerative disorders termed the GM2 gangliosidoses. The enzyme is composed of two subunits, alpha and beta. This gene encodes alpha subunit of beta-hexosaminidase. Western blot with this antibody raised against HEXA detected 60 kDa (precursor) and 54 kDa (mature) size bands (PMID: 27682588, PMID: 30341570).
Spécification
Spécification
HEXA
Polyclonal
Unconjugated
150 μL
3073
Liquid
Western Blot, Immunohistochemistry, ELISA
700 μg/mL