Learn More
Histone Deacetylase 2/HDAC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marque:  Novus Biologicals NBP1-87109
Code nomenclature Nacres: NA.46
Code produit. 18735923
  • 456.00€ / 0.10mL
Description
Histone Deacetylase 2/HDAC2 Polyclonal specifically detects Histone Deacetylase 2/HDAC2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Western Blot, Immunohistochemistry, Immunofluorescence
HDAC2
55 kDa
0.1 mL
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50, Knockdown Validated
This antibody was developed against Recombinant Protein corresponding to amino acids:IACDEEFSDSEDEGEGGRRNVADHKKGAKKARIEEDKKETEDKKTDVKEEDKSKDNSGEKTDTKGTKSEQLSNP
Affinity Purified
For Research Use Only