Learn More
Histone H3, Trimethyl Lys9 Antibody (6F12-H4), Novus Biologicals™
Code produit. 18715383 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.025 mL
0.1 mL
Conditionnement:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantité unitSize
18715383 0.025 mL 25µL
18731484 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit. 18715383 Fournisseur Novus Biologicals Code fournisseur NBP130141SS
Mouse Monoclonal Antibody has been used in 19 publications
Histone H3 Monoclonal specifically detects Histone H3 in Human, Mouse, Rat, Porcine, C. elegans, Drosophila, Invertebrate, Mammal, Yeast samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Spécification
Antigène Histone H3 (Trimethyl Lys9)
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 6F12-H4
Concentration 1 mg/mL
Conjugué Unconjugated
Dilution Western Blot 1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:1000. Use reported in scientific literature (PMID 23872316), Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 28270554), Immunohistochemistry-Paraffin 1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Numéro d’ordre du gène P84243, P84244
Alias de gène H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, H3K9Me3, HIST2H3C, histone H3.3, MGC87782, MGC87783
Symboles de gène(s) H3C14
Espèces hôtes Mouse
Immunogène This Histone H3 [Trimethyl Lys9] antibody (6F12-H4) was raised against a synthetic peptide made to an N-terminal region of Histone H3 (between amino acids 1-50). [UniProt# P84243]
Poids moléculaire de l’antigène 15 kDa
Méthode de purification Protein G purified
Quantité 0.025 mL
État réglementaire RUO
Disciplines de recherche Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primaire ou secondaire Primary
Identification génétique (Entrez) 126961
Spécificité du test All mammals including human, mouse and rat, Drosophila, C. elegans, yeast. Invertebrate/Blattella germanica (German cockroach) reactivity reported in scientific literature (PMID: 23872316).
Espèces cibles Human, Mouse, Rat, Pig, C. elegans, Drosophila, Invertebrate, Mammalia, Yeast
Contenu et stockage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Forme Purified
Isotype IgG1 κ
Afficher plus Afficher moins
For Research Use Only