missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGCS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17643-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This enzyme condenses acetyl-CoA with acetoacetyl-CoA to form HMG-CoA, which is the substrate for HMG-CoA reductase.Spécification
HMGCS1 | |
Polyclonal | |
Unconjugated | |
HMGCS1 | |
HMG CoA synthase, HMGCS, HMGCS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
208715, 29637, 3157 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P17425, Q01581, Q8JZK9 | |
HMGCS1 | |
HMGCS1 Fusion Protein Ag11825 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |