missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | HNF1A |
---|---|
Concentration | 0.56 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16811935
|
Proteintech
22426-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 29-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16801935
|
Proteintech
22426-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 29-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
This gene is expressed ubiquitously with higher levels in fetal than in adult tissues. It encodes a protein sharing 93% sequence identity with the mouse protein. Wolf-Hirschhorn syndrome (WHS) is a malformation syndrome associated with a hemizygous deletion of the distal short arm of chromosome 4. This gene is mapped to the 165 kb WHS critical region, and may play a role in the phenotype of the WHS or Pitt-Rogers-Danks syndrome. The encoded protein is found to be capable of reacting with HLA-A2-restricted and tumor-specific cytotoxic T lymphocytes, suggesting a target for use in specific immunotherapy for a large number of cancer patients. This protein has also been shown to be a member of the NELF (negative elongation factor) protein complex that participates in the regulation of RNA polymerase II transcription elongation.Spécification
HNF1A | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
P15257, P20823, P22361 | |
21405, 24817, 6927 | |
HNF1A Fusion Protein Ag18188 | |
Primary | |
-20°C | |
HNF1A |
0.56 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HNF 1 alpha, HNF 1A, HNF1, HNF1 homeobox A, HNF1A, LFB1, MODY3, TCF 1, TCF1, Transcription factor 1 | |
HNF1A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |