missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA2 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25044-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.Spécification
HOXA2 | |
Polyclonal | |
Unconjugated | |
HOXA2 | |
homeobox A2, Homeobox protein Hox 1K, Homeobox protein Hox A2, HOX1K, HOXA2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
103690123, 3199 | |
-20°C | |
Liquid |
Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43364, P31246 | |
HOXA2 | |
HOXA2 Fusion Protein Ag19224 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |