missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPSE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21128-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a heparanase enzyme. The encoded protein is a endoglycosidase that degrades heparin sulfate proteoglycans located on the extracellular matrix and cell surface. This protein may be involved in biological processes involving remodeling of the extracellular matrix including angiogenesis and tumor progression. Alternate splicing results in multiple transcript variants.Spécification
HPSE2 | |
Polyclonal | |
Unconjugated | |
HPSE2 | |
heparanase 2, HPA2, HPR2, HPSE2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
368128, 545291, 60495 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B2RY83, Q8WWQ2 | |
Hpse2 | |
HPSE2 Fusion Protein Ag15526 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |