missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRPT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12310-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
U2 auxiliary factor (U2AF), comprised of a large and a small subunit, is a non-snRNP protein required for the binding of U2 snRNP to the pre-mRNA branch site. This gene encodes the U2AF large subunit which contains a sequence-specific RNA-binding region with 3 RNA recognition motifs and an Arg/Ser-rich domain necessary for splicing. The large subunit binds to the polypyrimidine tract of introns early during spliceosome assembly. Multiple transcript variants have been detected for this gene, but the full-length natures of only two have been determined to date.Spécification
HRPT2 | |
Polyclonal | |
Unconjugated | |
CDC73 | |
C1orf28, CDC73, HPT JT, HRPT2, Hyperparathyroidism 2 protein, Parafibromin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
214498, 304832, 79577 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4V8C8, Q6P1J9, Q8JZM7 | |
CDC73 | |
HRPT2 Fusion Protein Ag2959 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |