missing translation for 'onlineSavingsMsg'
Learn More

HSPBAP1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 27631-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16835745

  • 399.00€ / 150µL
Expédition estimée: 03-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

HSPBAP1 (HSPB (heat shock 27kDa) associated protein 1), also known as PASS1 (protein associated with small stress protein 1), is a 488 amino acid cytoplasmic protein that contains one JMJC (Jumonji C) domain and shares 80% identity with its rat homolog, PASS1. Widely expressed with highest expression in ovary, thymus and pancreas, HSPBAP1 is thought to play a role in mediating cellular stress responses within the cell. Due to the presence of a JMJC domain, HSPBAP1 may be involved in chromatin remodeling events. Defects or translocations in the gene encoding HSPBAP1 are associated with renal cell carcinoma 1 (RCC1), suggesting a possible role for HSPBAP1 in carcinogenesis. Three isoforms of HSPBAP1 exist due to alternative splicing events.
Spécification

Spécification

HSPBAP1
Polyclonal
Unconjugated
HSPBAP1
HSPB1 associated protein 1, HSPBAP1, PASS1
Rabbit
Antigen Affinity Chromatography
RUO
79663
-20°C
Liquid
Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96EW2
HSPBAP1
HSPBAP1 Fusion Protein Ag26428
150 μL
Primary
Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions