missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPBAP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27631-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
HSPBAP1 (HSPB (heat shock 27kDa) associated protein 1), also known as PASS1 (protein associated with small stress protein 1), is a 488 amino acid cytoplasmic protein that contains one JMJC (Jumonji C) domain and shares 80% identity with its rat homolog, PASS1. Widely expressed with highest expression in ovary, thymus and pancreas, HSPBAP1 is thought to play a role in mediating cellular stress responses within the cell. Due to the presence of a JMJC domain, HSPBAP1 may be involved in chromatin remodeling events. Defects or translocations in the gene encoding HSPBAP1 are associated with renal cell carcinoma 1 (RCC1), suggesting a possible role for HSPBAP1 in carcinogenesis. Three isoforms of HSPBAP1 exist due to alternative splicing events.Spécification
HSPBAP1 | |
Polyclonal | |
Unconjugated | |
HSPBAP1 | |
HSPB1 associated protein 1, HSPBAP1, PASS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79663 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96EW2 | |
HSPBAP1 | |
HSPBAP1 Fusion Protein Ag26428 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |