missing translation for 'onlineSavingsMsg'
Learn More

HTR1A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 55302-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16836795

  • 121.38€ / 20µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity.
Spécification

Spécification

HTR1A
Polyclonal
Unconjugated
HTR1A
5 HT 1A, 5 HT1A, 5HT1a, ADRB2RL1, ADRBRL1, G 21, HTR1A, Serotonin receptor 1A
Rabbit
Antigen Affinity Chromatography
RUO
3350
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08908
HTR1A
Peptide
20 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions