missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human B7-H7/HHLA2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Marque: R&D Systems AF8084-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
B7-H7/HHLA2 Polyclonal specifically detects B7-H7/HHLA2 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Spécification
| B7-H7/HHLA2 | |
| Polyclonal | |
| Western Blot 1 ug/mL, Immunocytochemistry 1-15 ug/mL | |
| Q9UM44 | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 11148 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| B7H7, HERV-H LTR-associating 2, HERV-H LTR-associating protein 2, HHLA2, Human endogenous retrovirus-H long terminal repeat-associating protein 2 | |
| Human embryonic kidney cell line HEK293-derived human B7-H7/HHLA2 Met1-Asn344 Accession # Q9UM44 | |
| 25 μg | |
| Primary | |
| Detects human B7-H7/HHLA2 in direct ELISAs. | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only (RUO)
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu