missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BDNF Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Marque: R&D Systems MAB248-100
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
BDNF Monoclonal specifically detects BDNF in Human samples. It is validated for Western Blot, Immunohistochemistry.
Spécification
| BDNF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 627 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 35928 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P23560 | |
| BDNF | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human BDNF Arg128-Arg247 Accession # P23560 | |
| 100 μg | |
| Primary | |
| Detects human BDNF in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 5% cross-reactivity with recombinant human (rh) beta-NGF and rr beta-NGF. | |
| Human | |
| Ascites |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu