missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CDCP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00€ - 396.00€
Especificaciones
| Antigène | CDCP1 |
|---|---|
| Clone | 309121 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation 25 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Immunoprecipitation, CyTOF |
| Código de producto | Marca | Quantité | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantité | Precio | Cantidad y disponibilidad | |||||
18782559
![]() |
R&D Systems
MAB26662-SP |
25 μg |
209.00€
25µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
18140831
|
R&D Systems
MAB26662 |
100 μg |
396.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Descripción
CDCP1 Monoclonal specifically detects CDCP1 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, CyTOF-ready.Especificaciones
| CDCP1 | |
| LYOPH | |
| Flow Cytometry, Immunoprecipitation, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_073753 | |
| 64866 | |
| Mouse myeloma cell line NS0-derived recombinant human CDCP1 isoform 1 Ala33-Leu666 Accession # NP_073753 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CDCP1 in direct ELISAs. Recognizes intact protein (aa 33-666) of human CDCP1. |
| 309121 | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation 25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD318, CD318 antigen, CUB domain containing protein 1, CUB domain-containing protein 1, Membrane glycoprotein gp140, SIMA135, SIMA135TRASK, Subtractive immunization M plus HEp3-associated 135 kDa protein, Transmembrane and associated with src kinases | |
| CDCP1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto