missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FCRN Antibody, R&D Systems™

Mouse Monoclonal Antibody
227.00€ - 541.00€
Spécification
| Antigène | FCRN/FCGRT |
|---|---|
| Clone | 937508 |
| Dilution | Western Blot 5 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
18777638
![]() |
R&D Systems
MAB8639-SP |
25 μg |
227.00€
25µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
18181181
|
R&D Systems
MAB8639-100 |
100 μg |
541.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
FCRN/FCGRT Monoclonal specifically detects FCRN/FCGRT in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.Spécification
| FCRN/FCGRT | |
| Western Blot 5 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FCGRT, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig | |
| Chinese hamster ovary cell line CHO-derived recombinant human FCRN Ala24-Ser297 Accession # P55899 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human FCRN in direct ELISAs. |
| 937508 | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| P55899 | |
| 2217 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit