missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IBSP/Sialoprotein II Antibody, R&D Systems™
Goat Polyclonal Antibody
Marque: R&D Systems AF4014
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IBSP/Sialoprotein II Polyclonal specifically detects IBSP/Sialoprotein II in Human samples. It is validated for Western Blot.
Spécification
| IBSP/Sialoprotein II | |
| Polyclonal | |
| Unconjugated | |
| AAC95490 | |
| IBSP | |
| Chinese hamster ovary cell line CHO-derived recombinant human IBSP/Sialoprotein II Phe17-Gln317 Accession # AAC95490 | |
| 100 μg | |
| Primary | |
| Detects human IBSP/Sialoprotein II in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| BNSP, Bone sialoprotein, Bone sialoprotein 2, BSP, BSP 2, BSP II, BSP2, BSPII, BSP-II, Cell binding sialoprotein, IBSP, Integrin binding sialoprotein, SP II, SPII, SP-II | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3381 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu