missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-alpha 2/IFNA2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
201.00€ - 417.00€
Spécification
| Antigène | IFN-alpha 2 |
|---|---|
| Clone | 967314 |
| Dilution | Immunocytochemistry 8-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
18796929
![]() |
R&D Systems
MAB9345-SP |
25 μg |
201.00€
25µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
18701939
![]() |
R&D Systems
MAB9345-100 |
100 μg |
417.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
IFN-alpha 2 Monoclonal specifically detects IFN-alpha 2 in Human samples. It is validated for Immunocytochemistry.Spécification
| IFN-alpha 2 | |
| Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P01563 | |
| 3440 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 967314 | |
| Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| alpha-2a interferon, Ifa2, IFNA, IFNA2, IFNA2a, IFNA2b, IFNA2c, IFNAA, IFNalpha 2, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765 | |
| Human embryonic kidney cell line HEK-293-derived transfected with human IFN-alpha 2/IFNA2, Met1-Glu188, Accession # P01563 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IFN-alpha 2/IFNA2 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IFNE, rhIFNB, rhIFNG, or recombinant cotton rat IFNA is observed. In sandwich ELISAs, less than 8% cross-reactivity with rhIFNA-7, rhIFNA-8, and rhIFNA-10 is observed. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit