missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MAO-A Antibody, R&D Systems™

Sheep Polyclonal Antibody
Marque: R&D Systems AF8247-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MAO-A Polyclonal specifically detects MAO-A in Human samples. It is validated for Western Blot.
Spécification
| MAO-A | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| amine oxidase [flavin-containing] A, EC 1.4.3, EC 1.4.3.4, MAOA, MAO-A, monoamine oxidase A, Monoamine oxidase type A | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4128 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.25 ug/mL | |
| P21397 | |
| MAOA | |
| S. frugiperda insect ovarian cell line Sf21-derived recombinant human MAO-A Met1-Ser497 (predicted) Accession # P21397 | |
| 25 μg | |
| Primary | |
| Detects human MAO-A in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu