missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse CD11b Rat, APC-Cy7, Clone: M1/70, enQuireBio™
Rat Monoclonal Antibody
Marque: EnquireBio QAB22-APC7-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Human/Mouse CD11b | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Protein A/G Purified | |
RUO | |
Store at 2-4°C. |
Flow Cytometry | |
M1/70 | |
APC-Cyanine7 | |
Rat | |
100 μg | |
Primary | |
IgG2b κ |