Learn More
Human/Mouse CD11b Rat, Qfluor 710, Clone: M1/70, enQuireBio™
Rat Monoclonal Antibody
Marque: EnquireBio QAB22-QF710-25ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Human/Mouse CD11b | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Protein A/G Purified | |
RUO | |
Store at 2-4°C. |
Flow Cytometry | |
M1/70 | |
Qfluor 710 | |
Rat | |
25 μg | |
Primary | |
IgG2b κ |