missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EnquireBio Human/Mouse CD45R / B220 Rat, Qfluor 710, Clone: RA3-6B2, enQuireBio™
Rat Monoclonal Antibody
Marque: EnquireBio QAB41-QF710-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
Human/Mouse CD45R / B220 | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Ptprc | |
Protein A/G Purified | |
RUO | |
19264 | |
IgG2a κ |
Flow Cytometry | |
RA3-6B2 | |
Qfluor 710 | |
NP_001255215 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |