missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MSPR/Ron Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Marque: R&D Systems AF691-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MSPR/Ron Polyclonal specifically detects MSPR/Ron in Human samples. It is validated for Western Blot, Immunoprecipitation, Blockade of Receptor-ligand Interaction.
Spécification
| MSPR/Ron | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD136, CD136 antigen, CDw136c-met-related tyrosine kinase, EC 2.7.10, EC 2.7.10.1, macrophage stimulating 1 receptor (c-met-related tyrosine kinase), MSP R, MSP receptor, MST1R, p185-Ron, Protein-tyrosine kinase 8, PTK8, PTK8 protein tyrosine kinase 8, Ron, RONmacrophage-stimulating protein receptor, soluble RON variant 1, soluble RON variant 2, soluble RON variant 3, soluble RON variant 4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4486 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunoprecipitation 1 ug/1 mg cell lysate, Blockade of Receptor-ligand Interaction 1-3 ug/mL | |
| CAA49634 | |
| MST1R | |
| Mouse myeloma cell line NS0-derived recombinant human MSP R Glu25-Ser956 Accession # CAA49634 | |
| 25 μg | |
| Primary | |
| Delects human MSP R/Ron in direct ELISAs and Western blots. In these formats, approximately 25% cross-reactivity with recombinant mouse MSP R is observed. | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu