missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MSPR/Ron Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
209.00€ - 506.00€
Spécification
| Antigène | MSPR/Ron |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 2 ug/mL, Immunoprecipitation 1 ug/1 mg cell lysate, Blockade of Receptor-ligand Interaction 1-3 ug/mL |
| Applications | Western Blot, Immunoprecipitation, Blockade of Receptor-ligand Interaction |
| Classification | Polyclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
18743299
![]() |
R&D Systems
AF691-SP |
25 μg |
209.00€
25µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
18048721
![]() |
R&D Systems
AF691 |
100 μg |
506.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
MSPR/Ron Polyclonal specifically detects MSPR/Ron in Human samples. It is validated for Western Blot, Immunoprecipitation, Blockade of Receptor-ligand Interaction.Spécification
| MSPR/Ron | |
| Western Blot 2 ug/mL, Immunoprecipitation 1 ug/1 mg cell lysate, Blockade of Receptor-ligand Interaction 1-3 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| CD136, CD136 antigen, CDw136c-met-related tyrosine kinase, EC 2.7.10, EC 2.7.10.1, macrophage stimulating 1 receptor (c-met-related tyrosine kinase), MSP R, MSP receptor, MST1R, p185-Ron, Protein-tyrosine kinase 8, PTK8, PTK8 protein tyrosine kinase 8, Ron, RONmacrophage-stimulating protein receptor, soluble RON variant 1, soluble RON variant 2, soluble RON variant 3, soluble RON variant 4 | |
| MST1R | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation, Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| RUO | |
| CAA49634 | |
| 4486 | |
| Mouse myeloma cell line NS0-derived recombinant human MSP R Glu25-Ser956 Accession # CAA49634 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Delects human MSP R/Ron in direct ELISAs and Western blots. In these formats, approximately 25% cross-reactivity with recombinant mouse MSP R is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title