missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neurogranin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marque: R&D Systems MAB79473-100
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Neurogranin Monoclonal antibody specifically detects Neurogranin in Human samples. It is validated for ELISA
Spécification
| Neurogranin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| calmodulin-binding protein, hng, neurogranin, neurogranin (protein kinase C substrate, RC3), Ng, NRGN, protein kinase C substrate, RC3, RC3ng | |
| E. coli-derived recombinant human Neurogranin, Met1-Asp78, Accession # Q92686 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 1034203 | |
| ELISA | |
| Q92686 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4900 | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu