missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate Angiopoietin-like Protein 4/ANGPTL4 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 9 publications
Marque: R&D Systems AF3485
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Angiopoietin-like Protein 4/ANGPTL4 Polyclonal specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Human, Primate samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Spécification
| Angiopoietin-like Protein 4/ANGPTL4 | |
| Polyclonal | |
| Unconjugated | |
| Q9BY76 | |
| ANGPTL4 | |
| Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-like 4/ANGPLT4, Leu165-Ser406, Accession # Q9BY76 | |
| 100 μg | |
| Primary | |
| Detects human ANGPTL4 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human (rh) ANGPTL3, rhAngiopoietin-1, -2, -3, and -4 is observed. In Western blots, approximately 1% cross-reactivity with rhANGPLT4 N-terminal Fragment is observed. | |
| Human, Primate | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 50 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Angiopoietin like Protein 4, angiopoietin-like 4, Angiopoietin-like protein 4, ANGPTL4, ARP4fasting-induced adipose factor, FIAF, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARP, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGAR, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 51129 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu