missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate Angiopoietin-like Protein 4/ANGPTL4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
227.00€ - 609.00€
Spécification
| Antigène | Angiopoietin-like Protein 4/ANGPTL4 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Simple Western 50 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
18710818
![]() |
R&D Systems
AF3485-SP |
25 μg |
227.00€
25µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
18001482
|
R&D Systems
AF3485 |
100 μg |
609.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Angiopoietin-like Protein 4/ANGPTL4 Polyclonal specifically detects Angiopoietin-like Protein 4/ANGPTL4 in Human, Primate samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Spécification
| Angiopoietin-like Protein 4/ANGPTL4 | |
| Western Blot 0.1 ug/mL, Simple Western 50 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Polyclonal | |
| Goat | |
| Human, Primate | |
| Angiopoietin like Protein 4, angiopoietin-like 4, Angiopoietin-like protein 4, ANGPTL4, ARP4fasting-induced adipose factor, FIAF, FIAFhepatic angiopoietin-related protein, Hepatic fibrinogen/angiopoietin-related protein, HFARP, HFARPANGPTL2, NL2, peroxisome proliferator-activated receptor (PPAR) gamma inducedangiopoietin-related protein, PGAR, PGARangiopoietin-related protein 4, pp1158, PPARG angiopoietin related protein | |
| ANGPTL4 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Q9BY76 | |
| 51129 | |
| Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-like 4/ANGPLT4, Leu165-Ser406, Accession # Q9BY76 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human ANGPTL4 in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human (rh) ANGPTL3, rhAngiopoietin-1, -2, -3, and -4 is observed. In Western blots, approximately 1% cross-reactivity with rhANGPLT4 N-terminal Fragment is observed. |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit