missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate IL-17F Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Marque: R&D Systems AF1335-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IL-17F Polyclonal specifically detects IL-17F in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Dual RNAscope ISH-IHC, ELISA Capture (Matched Antibody Pair).
Spécification
| IL-17F | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cytokine ML-1, IL17F, IL-17Finterleukin-17F, interleukin 17F | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 112744 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, In Situ Hybridization (ISH), ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL, Immunocytochemistry 5-15 ug/mL, Dual RNAscope ISH-IHC 3-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| AAK83350 | |
| IL17F | |
| E. coli-derived recombinant human IL-17F (R&D Systems, Catalog # 1335-IL) Arg31-Gln163 Accession # AAK83350 | |
| 25 μg | |
| Primary | |
| Detects human IL-17F in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity recombinant human (rh) IL-17, rhIL-17B, rhIL-17C, rhIL-17D, rhIL-17E, and recombinant mouse IL-17F is observed. | |
| Human, Primate | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu