missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate IL-17F Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
214.00€ - 671.00€
Spécification
| Antigène | IL-17F |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL, Immunocytochemistry 5-15 ug/mL, Dual RNAscope ISH-IHC 3-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, In Situ Hybridization (ISH), ELISA |
| Classification | Polyclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
18787887
![]() |
R&D Systems
AF1335-SP |
25 μg |
214.00€
25µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
18011862
![]() |
R&D Systems
AF1335 |
100 μg |
671.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Beskrivning
IL-17F Polyclonal specifically detects IL-17F in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Dual RNAscope ISH-IHC, ELISA Capture (Matched Antibody Pair).Specifikationer
| IL-17F | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL, Immunocytochemistry 5-15 ug/mL, Dual RNAscope ISH-IHC 3-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Polyclonal | |
| Goat | |
| Human, Primate | |
| Cytokine ML-1, IL17F, IL-17Finterleukin-17F, interleukin 17F | |
| IL17F | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, In Situ Hybridization (ISH), ELISA | |
| Unconjugated | |
| RUO | |
| AAK83350 | |
| 112744 | |
| E. coli-derived recombinant human IL-17F (R&D Systems, Catalog # 1335-IL) Arg31-Gln163 Accession # AAK83350 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human IL-17F in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity recombinant human (rh) IL-17, rhIL-17B, rhIL-17C, rhIL-17D, rhIL-17E, and recombinant mouse IL-17F is observed. |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel