missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SUSD2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Marque: R&D Systems MAB90562-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SUSD2 Monoclonal specifically detects SUSD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Spécification
| SUSD2 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| BK65A6.2, FLJ22778, Sushi domain (SCR repeat) containing, sushi domain containing 2, sushi domain-containing protein 2 | |
| HEK293 human embryonic kidney cell line-derived human SUSD2 Met1-Ala785 Accession # Q9UGT4 | |
| 25 μg | |
| Primary | |
| Detects human SUSD2 in direct ELISAs and Western blots. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| 1279A | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 3-25 ug/mL | |
| Q9UGT4 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 56241 | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu