missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Tripeptidyl-Peptidase I/TPP1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Marque: R&D Systems AF2237
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Tripeptidyl-Peptidase I/TPP1 Polyclonal specifically detects Tripeptidyl-Peptidase I/TPP1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Spécification
| Tripeptidyl-Peptidase I/TPP1 | |
| Polyclonal | |
| Unconjugated | |
| O14773 | |
| TPP1 | |
| Mouse myeloma cell line NS0-derived recombinant human Tripeptidyl-Peptidase I/TPP1 Ser20-Pro563 Accession # O14773 | |
| 100 μg | |
| Primary | |
| Detects human Tripeptidyl-Peptidase I/TPP1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Cell growth-inhibiting gene 1 protein, ceroid-lipofuscinosis, neuronal 2, late infantile (Jansky-Bielschowsky disease), CLN2, CLN2EC 3.4.14.9, growth-inhibiting protein 1, LINCL, LPIC, lysosomal pepstatin insensitive protease, Lysosomal pepstatin-insensitive protease, MGC21297, TPP1, TPP-1, TPP-I, Tripeptidyl aminopeptidase, tripeptidyl peptidase I, tripeptidyl-peptidase 1, Tripeptidyl-peptidase I, TripeptidylPeptidase I | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1200 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu