missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Tripeptidyl-Peptidase I/TPP1 Antibody, R&D Systems™

Goat Polyclonal Antibody
209.00€ - 501.00€
Spécification
| Antigène | Tripeptidyl-Peptidase I/TPP1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
18706647
![]() |
R&D Systems
AF2237-SP |
25 μg |
209.00€
25µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
18080582
![]() |
R&D Systems
AF2237 |
100 μg |
501.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Tripeptidyl-Peptidase I/TPP1 Polyclonal specifically detects Tripeptidyl-Peptidase I/TPP1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Spécification
| Tripeptidyl-Peptidase I/TPP1 | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| Cell growth-inhibiting gene 1 protein, ceroid-lipofuscinosis, neuronal 2, late infantile (Jansky-Bielschowsky disease), CLN2, CLN2EC 3.4.14.9, growth-inhibiting protein 1, LINCL, LPIC, lysosomal pepstatin insensitive protease, Lysosomal pepstatin-insensitive protease, MGC21297, TPP1, TPP-1, TPP-I, Tripeptidyl aminopeptidase, tripeptidyl peptidase I, tripeptidyl-peptidase 1, Tripeptidyl-peptidase I, TripeptidylPeptidase I | |
| TPP1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| O14773 | |
| 1200 | |
| Mouse myeloma cell line NS0-derived recombinant human Tripeptidyl-Peptidase I/TPP1 Ser20-Pro563 Accession # O14773 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Tripeptidyl-Peptidase I/TPP1 in direct ELISAs and Western blots. |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit