missing translation for 'onlineSavingsMsg'
Learn More

ICOS Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13338-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16831494

  • 325.38€ / 150µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

13338-1-AP recognizes two bands of 25-30 kDa and 45-50 kDa, and the additional 45-50 kDa band may be due to surface disulfide-linked homodimeric glycoprotein.

Inducible T-cell costimulator is an immune checkpoint protein that in humans is encoded by the ICOS gene. The protein encoded by this gene belongs to the CD28 and CTLA-4 cell-surface receptor family. It forms homodimers and plays an important role in cell-cell signaling, immune responses, and regulation of cell proliferation.
Spécification

Spécification

ICOS
Polyclonal
Unconjugated
ICOS
AILIM, CD278, ICOS, inducible T cell co stimulator, Inducible T cell costimulator
Rabbit
Antigen Affinity Chromatography
RUO
29851
-20°C
Liquid
Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y6W8
ICOS
ICOS Fusion Protein Ag4155
150 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions