Learn More
ICOSLG Mouse anti-Human, Unconjugated, Clone: 2G1F8, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67003-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16829365
  • 121.38€ / 20µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 
Description
Description
The ICOSLG antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ICOSLG. This antibody recognizes human antigen. The ICOSLG antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
ICOSLG
Monoclonal
1000 μg/mL
Mouse
Primary
Human
ELISA, Western Blot
2G1F8
Unconjugated
20 μL
23308
IgG2a