missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN gamma Rat, APC, Clone: XMG1.2, enQuireBio™
Rat Monoclonal Antibody
Marque: EnquireBio QAB83-APC-100ug
Informations supplémentaires : Poids : 0.01000kg
Spezifikation
IFN gamma | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
Ifng | |
Protein A/G Purified | |
RUO | |
15978 | |
IgG1 κ |
Flow Cytometry | |
XMG1.2 | |
APC | |
NP_032363 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |